4'-Brom-3'-chloracetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Bromo-3-chlorophenyl)ethanone
OR301142-10G
OR301142-1G
OR301142-5G
APOSOR301142-10GEA
132
EUR
APOSOR301142-10G
APOSOR301142-1G
APOSOR301142-5G
4'-Brom-3'-chloracetophenon 98%
4'-Brom-3'-chloracetophenon
Formel:
C₈H₆BrClO Molecular Weight: 233,4900 g/mol Schmelzpunkt: 80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14636445 CAS-Nummer: 3114-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...