6-Methoxy-1,2,3,4-tetrahydroisochinolin

Lieferant: Apollo Scientific
Achtung

OR301147-5G
APOSOR301147-5GEA 194 EUR
APOSOR301147-5G
6-Methoxy-1,2,3,4-tetrahydroisochinolin
6-Methoxy-1,2,3,4-tetrahydroisochinolin
Formel: C₁₀H₁₃NO
Molecular Weight: 163,2163 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01717129
CAS-Nummer: 42923-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR