(3-Methyloxetan-3-yl)methanamine

Lieferant: Apollo Scientific

OR301149-5G
APOSOR301149-5GEA 84.1 EUR
APOSOR301149-5G
(3-Methyloxetan-3-yl)methanamine
(3-Methyloxetan-3-yl)methanamine
Formel: C₅H₁₁NO
Molecular Weight: 101,1469 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703640
CAS-Nummer: 153209-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR