7-Bromo-3,4-dihydro-2H-isoquinolin-1-one 98%

Lieferant: Apollo Scientific
Achtung

OR301150-1G OR301150-250MG OR301150-5G
APOSOR301150-1GEA 102 EUR
APOSOR301150-1G APOSOR301150-250MG APOSOR301150-5G
7-Bromo-3,4-dihydro-2H-isoquinolin-1-one 98%
7-Bromo-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₉H₈BrNO
Molecular Weight: 226,0700 g/mol
Schmelzpunkt: 156…159 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08437642
CAS-Nummer: 891782-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR