2-Amino-6-methoxypyrazin

Lieferant: Apollo Scientific
Achtung

OR301157-5G
APOSOR301157-5GEA 202 EUR
APOSOR301157-5G
2-Amino-6-methoxypyrazin
2-Amino-6-methoxypyrazine
Formel: C₅H₇N₃O
Molecular Weight: 125,13 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09909689
CAS-Nummer: 6905-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR