3-Ethyl-2-iodtoluol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Ethyl-2-iod-3-methylbenzol, 2-Ethyl-6-methyliodbenzol

OR30116-10G OR30116-25G OR30116-1G OR30116-5G OR30116-50G
APOSOR30116-10GEA 111 EUR
APOSOR30116-10G APOSOR30116-25G APOSOR30116-1G APOSOR30116-5G APOSOR30116-50G
3-Ethyl-2-iodtoluol
3-Ethyl-2-iodtoluol
Formel: C₉H₁₁I
Molecular Weight: 246.09 g/mol
Siedepunkt: 118…120 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052855
CAS-Nummer: 175277-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR