5-Methylpicolinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methylpyridine-2-carboxylic acid

OR301172-1G OR301172-5G
APOSOR301172-1GEA 0 EUR
APOSOR301172-1G APOSOR301172-5G
5-Methylpicolinsäure
5-Methylpicolinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137.1360 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01704358
CAS-Nummer: 4434-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR