Methyl indolizine-7-carboxylate

Lieferant: Apollo Scientific

OR301176-1G OR301176-5G
APOSOR301176-1GEA 255 EUR
APOSOR301176-1G APOSOR301176-5G
Methyl indolizine-7-carboxylate
Methyl indolizine-7-carboxylate
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10566763
CAS-Nummer: 887602-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR