2-Ethyl-6-methylbenzonitrile

Lieferant: Apollo Scientific
Achtung

OR30118-1G OR30118-250MG OR30118-500MG
APOSOR30118-1GEA 275 EUR
APOSOR30118-1G APOSOR30118-250MG APOSOR30118-500MG
2-Ethyl-6-methylbenzonitrile
2-Ethyl-6-methylbenzonitrile
Formel: C₁₀H₁₁N
Lagertemperatur: Kühlschrank
MDL: MFCD00052864
CAS-Nummer: 95881-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR