2-Brom-6-chlorpyrazin 98%
Lieferant: Apollo Scientific
OR301196-1G
OR301196-5G
APOSOR301196-1GEA
130
EUR
APOSOR301196-1G
APOSOR301196-5G
2-Brom-6-chlorpyrazin 98%
2-Bromo-6-chloropyrazine
Formel:
C₄H₂BrClN₂ Molecular Weight: 193,4291 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08752648 CAS-Nummer: 916791-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...