5-Hydroxynicotinonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR301211-5G
APOSOR301211-5GEA
55.1
EUR
APOSOR301211-5G
5-Hydroxynicotinonitrile 97%
5-Hydroxynicotinonitrile
Formel:
C6H4N2O Molecular Weight: 120,11 g/mol Schmelzpunkt: 213…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044307 CAS-Nummer: 152803-24-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...