2-(Chlormethyl)oxazol

Lieferant: Apollo Scientific
Achtung

OR301217-1G OR301217-250MG
APOSOR301217-1GEA 261 EUR
APOSOR301217-1G APOSOR301217-250MG
2-(Chlormethyl)oxazol
2-(Chloromethyl)oxazole
Formel: C₄H₄ClNO
Molecular Weight: 117,53 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD04114926
CAS-Nummer: 185246-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR