6-Bromo-1H-pyrrolo[3,2-b]pyridine 97%
Lieferant: Apollo Scientific
OR301260-25G
APOSOR301260-25GEA
137
EUR
APOSOR301260-25G
6-Bromo-1H-pyrrolo[3,2-b]pyridine 97%
6-Bromo-1H-pyrrolo[3,2-b]pyridine
Formel:
C₇H₅BrN₂ Molecular Weight: 197,0300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09266227 CAS-Nummer: 944937-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...