4-Chlor-2-iodanisol

Lieferant: Apollo Scientific

OR30128-100G OR30128-25G
APOSOR30128-100GEA 0 EUR
APOSOR30128-100G APOSOR30128-25G
4-Chlor-2-iodanisol
Formel: C₇H₆ClIO
Siedepunkt: 133…135 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052858
CAS-Nummer: 52807-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR