2-Chloro-3,5-diiodo-4-pyridinamine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-3,5-diiodopyridin-4-amine

OR301296-1G OR301296-5G
APOSOR301296-1GEA 270 EUR
APOSOR301296-1G APOSOR301296-5G
2-Chloro-3,5-diiodo-4-pyridinamine 95%
2-Chloro-3,5-diiodo-4-pyridinamine
Formel: C₅H₃ClI₂N₂
Molecular Weight: 380.35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12498707
CAS-Nummer: 1171919-00-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR