5,6-Dichloro-2,3-dihydro-1H-inden-1-one 98%

Lieferant: Apollo Scientific

Synonyme: 5,6-Dichloro-indanone

OR301317-5G
APOSOR301317-5GEA 105 EUR
APOSOR301317-5G
5,6-Dichloro-2,3-dihydro-1H-inden-1-one 98%
5,6-Dichloro-2,3-dihydro-1H-inden-1-one
Formel: C₉H₆Cl₂O
Molecular Weight: 201,0493 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09032944
CAS-Nummer: 68755-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR