Methyl-2-thiophencarboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Methyl Ester
OR30134-1G
OR30134-25G
OR30134-5G
APOSOR30134-1GEA
50.9
EUR
APOSOR30134-1G
APOSOR30134-25G
APOSOR30134-5G
Methyl-2-thiophencarboxylat 98%
Methyl-2-thiophencarboxylat
Formel:
C₆H₆O₂S Molecular Weight: 142,17 g/mol Siedepunkt: 94…96 °C/14mmHg Dichte: 1,23 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016895 CAS-Nummer: 5380-42-7 EINECS: 226-371-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...