Methyl-3-amino-4-methoxybenzoat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-4-methoxybenzoesäuremethylester
OR30135-5G
APOSOR30135-5GEA
68.3
EUR
APOSOR30135-5G
Methyl-3-amino-4-methoxybenzoat 97%
Methyl-3-amino-4-methoxybenzoat
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Schmelzpunkt: 82…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017196 CAS-Nummer: 24812-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...