3-Methyloxetane-3-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl-3-oxetanecarboxylic acid

OR301351-5G OR301351-25G
APOSOR301351-5GEA 56.7 EUR
APOSOR301351-5G APOSOR301351-25G
3-Methyloxetane-3-carboxylic acid 98%
3-Methyloxetane-3-carboxylic acid
Formel: C₅H₈O₃
Molecular Weight: 116.116 g/mol
Schmelzpunkt: 58…63 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09842435
CAS-Nummer: 28562-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR