Benzyl(4-oxocyclohexyl)carbamate 97%

Lieferant: Apollo Scientific

Synonyme: Benzyl (4-oxocyclohexyl)carbamate, 4-N-Cbz-aminocyclohexanone

OR301374-100G OR301374-25G OR301374-5G
APOSOR301374-100GEA 333 EUR
APOSOR301374-100G APOSOR301374-25G APOSOR301374-5G
Benzyl(4-oxocyclohexyl)carbamate 97%
Benzyl(4-oxocyclohexyl)carbamate
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658393
CAS-Nummer: 16801-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR