(4-Bromphenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

OR30163-100G OR30163-25G
APOSOR30163-100GEA 29 EUR
APOSOR30163-100G APOSOR30163-25G
(4-Bromphenyl)essigsäure 97%
(4-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 117…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004342
CAS-Nummer: 1878-68-8
EINECS: 217-523-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR