3,4,5-Trimethoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4,5-Trimethoxybenzeneboronic acid, Dihydroxy(3,4,5-trimethoxyphenyl)borane, (3,4,5-Trimethoxyphenyl)metaboric acid

OR3017-25G OR3017-5G OR3017-1G
APOSOR3017-25GEA 231 EUR
APOSOR3017-25G APOSOR3017-5G APOSOR3017-1G
3,4,5-Trimethoxyphenylboronsäure 98%
3,4,5-Trimethoxyphenylboronsäure
Formel: C₉H₁₃BO₅
Molecular Weight: 212,01 g/mol
Schmelzpunkt: 244…247 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075676
CAS-Nummer: 182163-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR