1-Methyl-4-nitro-1H-pyrazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-4-nitropyrazole
OR302024-100G
APOSOR302024-100GEA
74.1
EUR
APOSOR302024-100G
1-Methyl-4-nitro-1H-pyrazole 98%
1-Methyl-4-nitro-1H-pyrazole
Formel:
C₄H₅N₃O₂ Molecular Weight: 127,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00464261 CAS-Nummer: 3994-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...