2-Hydrazinopyrimidine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydrazinylpyrimidine

OR302061-1G OR302061-5G OR302061-25G
APOSOR302061-1GEA 21.9 EUR
APOSOR302061-1G APOSOR302061-5G APOSOR302061-25G
2-Hydrazinopyrimidine 97%
2-Hydrazinopyrimidine
Formel: C₄H₆N₄
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 110…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01249337
CAS-Nummer: 7504-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR