3-Amino-5-phenyl-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-5-phenylpyrazole
OR3021-1G
OR3021-5G
OR3021-10G
APOSOR3021-1GEA
58
EUR
APOSOR3021-1G
APOSOR3021-5G
APOSOR3021-10G
3-Amino-5-phenyl-1H-pyrazol
3-Amino-5-phenyl-1H-pyrazole
Formel:
C₉H₉N₃ Molecular Weight: 159.19 g/mol Schmelzpunkt: 124…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191749 CAS-Nummer: 1572-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...