Methyl-5-brompyrazin-2-carboxylat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-bromopyrazine-2-carboxylate
OR302107-100MG
OR302107-1G
OR302107-5G
APOSOR302107-100MGEA
72.5
EUR
APOSOR302107-100MG
APOSOR302107-1G
APOSOR302107-5G
Methyl-5-brompyrazin-2-carboxylat 98%
Methyl-5-brompyrazin-2-carboxylat
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217.02 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09991715 CAS-Nummer: 210037-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...