4-Bromo-2-chlorothiazole

Lieferant: Apollo Scientific
Gefahr

OR302112-1G OR302112-5G
APOSOR302112-1GEA 348 EUR
APOSOR302112-1G APOSOR302112-5G
4-Bromo-2-chlorothiazole
4-Bromo-2-chlorothiazole
Formel: C₃HBrClNS
Molecular Weight: 198.47 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09746336
CAS-Nummer: 92977-45-2
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR