1-Methyl-1H-pyrazol-5-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1-Methyl-1H-pyrazol-5-yl)boronic acid, (1-Methyl-1H-pyrazol-5-yl)-boronic acid
OR302125-10G
OR302125-25G
APOSOR302125-10GEA
79.9
EUR
APOSOR302125-10G
APOSOR302125-25G
1-Methyl-1H-pyrazol-5-boronsäure
1-Methyl-1H-pyrazol-5-boronsäure
Formel:
C₄H₇BN₂O₂ Molecular Weight: 125.92 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11053031 CAS-Nummer: 720702-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...