1-Boc-1H-pyrazol-5-boronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1-(tert-Butoxycarbonyl)-1H-pyrazol-5-yl)boronic acid, 1H-Pyrazole-5-boronic acid, N1-BOC protected, 1-Boc-1H-pyrazole-5-boronic Acid
OR302126-1G
OR302126-5G
APOSOR302126-1GEA
105
EUR
APOSOR302126-1G
APOSOR302126-5G
1-Boc-1H-pyrazol-5-boronsäure 95%
1-Boc-1H-pyrazole-5-boronic acid
Formel:
C₈H₁₃BN₂O₄ Molecular Weight: 212.0108 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD12025961 CAS-Nummer: 1217500-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...