1-(4-Bromophenyl)-2-hydroxy-1-ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Bromo-2-hydroxyacetophenone

OR30215-1G OR30215-5G OR30215-250MG
APOSOR30215-1GEA 109 EUR
APOSOR30215-1G APOSOR30215-5G APOSOR30215-250MG
1-(4-Bromophenyl)-2-hydroxy-1-ethanone
1-(4-Bromophenyl)-2-hydroxy-1-ethanone
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 137…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174275
CAS-Nummer: 3343-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR