6-Methoxy-1,3-benzothiazol-2-carbonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxy-2-benzothiazolecarbonitrile, 6-Methoxybenzo[d]thiazole-2-carbonitrile

OR302169-5G
APOSOR302169-5GEA 95.7 EUR
APOSOR302169-5G
6-Methoxy-1,3-benzothiazol-2-carbonitril
2-Cyano-6-methoxybenzothiazole
Formel: C₉H₆N₂OS
Molecular Weight: 190,22 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010537
CAS-Nummer: 943-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR