6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid 97%

Lieferant: Apollo Scientific

OR302171-1G OR302171-250MG
APOSOR302171-1GEA 2290 EUR
APOSOR302171-1G APOSOR302171-250MG
6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid 97%
6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid
Formel: C₁₃H₁₇N₃O₄
Molecular Weight: 279,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973511
CAS-Nummer: 264623-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR