3-Chloro-1-(2,5-dimethoxyphenyl)propan-1-one

Lieferant: Apollo Scientific

OR30226-1G OR30226-250MG OR30226-500MG
APOSOR30226-1GEA 589 EUR
APOSOR30226-1G APOSOR30226-250MG APOSOR30226-500MG
3-Chloro-1-(2,5-dimethoxyphenyl)propan-1-one
3-Chloro-1-(2,5-dimethoxyphenyl)propan-1-one
Formel: C₁₁H₁₃ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00126487
CAS-Nummer: 50786-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR