3-Amino-1-methylpyridin-2(1H)-one 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR302360-1G
OR302360-250MG
APOSOR302360-1GEA
375
EUR
APOSOR302360-1G
APOSOR302360-250MG
3-Amino-1-methylpyridin-2(1H)-one 95%
3-Amino-1-methylpyridin-2(1H)-one
Formel:
C₆H₈N₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD12402373 CAS-Nummer: 33631-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...