4-Amino-1-methylpyridin-2(1H)-one 97%

Lieferant: Apollo Scientific
Achtung

OR302362-1G OR302362-250MG OR302362-5G
APOSOR302362-1GEA 224 EUR
APOSOR302362-1G APOSOR302362-250MG APOSOR302362-5G
4-Amino-1-methylpyridin-2(1H)-one 97%
4-Amino-1-methylpyridin-2(1H)-one
Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09832890
CAS-Nummer: 952182-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR