1-(Cyanacetyl)piperidin
Lieferant: Apollo Scientific
OR30243-25G
OR30243-5G
APOSOR30243-25GEA
98.8
EUR
APOSOR30243-25G
APOSOR30243-5G
1-(Cyanacetyl)piperidin
1-(Cyanacetyl)piperidin
Formel:
C₈H₁₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006487 CAS-Nummer: 15029-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...