2-Azabicyclo[2.2.1]heptane 97%
Lieferant: Apollo Scientific
OR302439-1G
OR302439-250MG
APOSOR302439-1GEA
319
EUR
APOSOR302439-1G
APOSOR302439-250MG
2-Azabicyclo[2.2.1]heptane 97%
2-Azabicyclo[2.2.1]heptane
Formel:
C₆H₁₁N Molecular Weight: 97,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864542 CAS-Nummer: 279-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...