1-Phenylcyclopropanamine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Phenylcyclopropylamine
OR302455-1G
OR302455-5G
APOSOR302455-1GEA
59.6
EUR
APOSOR302455-1G
APOSOR302455-5G
1-Phenylcyclopropanamine 95%
1-Phenylcyclopropanamine
Formel:
C₉H₁₁N Molecular Weight: 133,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863520 CAS-Nummer: 41049-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...