Methyl-6-aminopyrazine-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-aminopyrazine-2-carboxylate
OR302497-1G
OR302497-250MG
OR302497-5G
APOSOR302497-1GEA
185
EUR
APOSOR302497-1G
APOSOR302497-250MG
APOSOR302497-5G
Methyl-6-aminopyrazine-2-carboxylate
Methyl-6-aminopyrazine-2-carboxylate
Formel:
C₆H₇N₃O₂ Molecular Weight: 153.1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697802 CAS-Nummer: 118853-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...