5-(4-Methyl-1-piperazinyl)-2-pyridinamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-4-(6-aminopyridin-3-yl)piperazine

OR302515-25G OR302515-5G OR302515-100G
APOSOR302515-25GEA 77 EUR
APOSOR302515-25G APOSOR302515-5G APOSOR302515-100G
5-(4-Methyl-1-piperazinyl)-2-pyridinamin
5-(4-Methylpiperazin-1-yl)pyridin-2-amine
Formel: C₁₀H₁₆N₄
Molecular Weight: 192,2608 g/mol
Schmelzpunkt: 150…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10009563
CAS-Nummer: 571189-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR