5-Morpholino-2-pyridinamin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-morpholinopyridine

OR302520-1G OR302520-100MG OR302520-250MG
APOSOR302520-1GEA 58 EUR
APOSOR302520-1G APOSOR302520-100MG APOSOR302520-250MG
5-Morpholino-2-pyridinamin 98%
5-Morpholino-2-pyridinamin
Formel: C₉H₁₃N₃O
Molecular Weight: 179,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09388785
CAS-Nummer: 571189-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR