Ethyl-2-amino-1H-imidazole-5-carboxylate 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-amino-1H-imidazole-5-carboxylate
OR302532-1G
OR302532-5G
APOSOR302532-1GEA
63.8
EUR
APOSOR302532-1G
APOSOR302532-5G
Ethyl-2-amino-1H-imidazole-5-carboxylate 97+%
Ethyl-2-amino-1H-imidazole-5-carboxylate
Formel:
C₆H₉N₃O₂ Molecular Weight: 155.1546 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864446 CAS-Nummer: 149520-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...