6-Amino-2-pyridincarbonitril 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Aminopicolinonitrile

OR302542-1G OR302542-250MG
APOSOR302542-1GEA 37.7 EUR
APOSOR302542-1G APOSOR302542-250MG
6-Amino-2-pyridincarbonitril 97%
6-Amino-2-pyridincarbonitril
Formel: C₆H₅N₃
Molecular Weight: 119.1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044252
CAS-Nummer: 370556-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR