Ethyl-5-aminopyrazine-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-aminopyrazine-2-carboxylate

OR302550-1G OR302550-25G OR302550-5G
APOSOR302550-1GEA 144 EUR
APOSOR302550-1G APOSOR302550-25G APOSOR302550-5G
Ethyl-5-aminopyrazine-2-carboxylate 95%
Ethyl-5-aminopyrazine-2-carboxylate
Formel: C₇H₉N₃O₂
Molecular Weight: 167,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437665
CAS-Nummer: 54013-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR