6-Chloro-4-pyridazinamine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloropyridazin-4-amine, 5-Amino-3-chloropyridazine

OR302554-1G OR302554-5G OR302554-250MG
APOSOR302554-1GEA 149 EUR
APOSOR302554-1G APOSOR302554-5G APOSOR302554-250MG
6-Chloro-4-pyridazinamine 98%
6-Chloro-4-pyridazinamine
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03085861
CAS-Nummer: 29049-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR