5-Amino-4-brom-5-phenyl-1H-pyrazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Amino-4-bromo-1H-pyrazol-3-yl)benzene, 4-Bromo-3-phenyl-1H-pyrazol-5-amine

OR3026-5G OR3026-10G
APOSOR3026-5GEA 226 EUR
APOSOR3026-5G APOSOR3026-10G
5-Amino-4-brom-5-phenyl-1H-pyrazol 95%
5-Amino-4-brom-5-phenyl-1H-pyrazol
Formel: C₉H₈BrN₃
Molecular Weight: 238.08 g/mol
Schmelzpunkt: 112…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067992
CAS-Nummer: 2845-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR