3,4-Dichlorbenzhydrazid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,4-Dichlorbenzoesäurehydrazid, 3,4-Dichlorbenzol-1-carbohydrazid, 3,4-Dichlorbenzohydrazid
OR30269-1G
OR30269-250MG
APOSOR30269-1GEA
43.5
EUR
APOSOR30269-1G
APOSOR30269-250MG
3,4-Dichlorbenzhydrazid 97%
3,4-Dichlorbenzhydrazid
Formel:
C₇H₆Cl₂N₂O Molecular Weight: 205,04 g/mol Schmelzpunkt: 154…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017057 CAS-Nummer: 28036-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...