3-(Benzyloxy)-4-methoxybenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Benzyloxy-4-methoxybenzaldehyde
OR30299-100G
OR30299-25G
OR30299-500G
APOSOR30299-100GEA
116
EUR
APOSOR30299-100G
APOSOR30299-25G
APOSOR30299-500G
3-(Benzyloxy)-4-methoxybenzaldehyd
3-(Benzyloxy)-4-methoxybenzaldehyd
Formel:
C₁₅H₁₄O₃ Molecular Weight: 242,27 g/mol Siedepunkt: 120 °C/5,3mmHg Schmelzpunkt: 60…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003386 CAS-Nummer: 6346-05-0 EINECS: 228-752-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...