2,6-Diamino-5-bromopyrimidin-4-ol

Lieferant: Apollo Scientific

Synonyme: 2,6-Diamino-5-bromopyrimidin-4(1H)-one

OR303016-1G OR303016-5G
APOSOR303016-1GEA 180 EUR
APOSOR303016-1G APOSOR303016-5G
2,6-Diamino-5-bromopyrimidin-4-ol
2,6-Diamino-5-bromopyrimidin-4-ol
Formel: C₄H₅BrN₄O
Molecular Weight: 205,02 g/mol
Schmelzpunkt: 256…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023262
CAS-Nummer: 6312-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR