2,6-Diamino-5-chloropyrimidin-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR303017-1G
OR303017-500MG
APOSOR303017-1GEA
270
EUR
APOSOR303017-1G
APOSOR303017-500MG
2,6-Diamino-5-chloropyrimidin-4-ol
2,6-Diamino-5-chloropyrimidin-4-ol
Formel:
C₄H₅ClN₄O Schmelzpunkt: 199…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22196157 CAS-Nummer: 71552-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...